Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC4 Monoclonal Antibody (3C7.D8.E11.F), Rockland™
SDP

Catalog No. 501990630
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0630 25 μL
50-199-0629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0630 Supplier Rockland Immunochemicals Supplier No. 200301GY2S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human MUC4 protein. A BLAST analysis was used to suggest cross reactivity with MUC4 from human based on 100% sequence homology with the immunogen. Reactivity with MUC4 from other sources is not known.

Nearly all mucosal surfaces are protected by mucin transmembrane glycoproteins, from bronchioles to corneas to reproductive tracts. Mucin 4 (MUC4) is membrane-bound and is important in protecting and lubricating. MUC4 expression is erroneous in many carcinomas and sarcomas, including fibromyoxoid sarcoma and pancreatic adenocarcinoma. Immunohistochemical staining for MUC4 has been discussed as a potential biomarker for detection of these cancers. MUC4 seems to be most predominantly affected in Asian diseases - Taiwanese infertility and endometriosis, as well as gallstone formation in Chinese men, are connected to MUC4, where similar diseases in other populations do not share these MUC4 mutations. Because MUC4 is so important in mucosal surface protection, it is not surprising that it has been implicated as failing in many metastatic cancer invasions.

Specifications

Antigen MUC4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C7.D8.E11.F
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MUC4
Gene Accession No. Q99102
Gene Alias 4933405I11Rik; Ascites sialoglycoprotein; Ascites sialoglycoprotein 1; Ascites sialoglycoprotein 2; ASGP; ASGP1; ASGP-1; ASGP-2; HSA276359; MUC4; MUC-4; mucin 4; mucin 4, ASGP; mucin 4, cell surface associated; mucin 4, tracheobronchial; Mucin4; Mucin-4; Mucin-4 alpha chain; Mucin-4 beta chain; Pancreatic adenocarcinoma mucin; pre-sialomucin complex; Psmc; sialomucin ascites sialoglycoprotein-1; sialomucin complex; Testis mucin; Tracheobronchial mucin
Gene Symbols MUC4
Host Species Mouse
Immunogen This protein A purified antibody was prepared from tissue culture supernate produced by repeated immunizations with a peptide corresponding to an internal portion of human Mucin-4.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4585
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.