Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5AC Antibody (2-11M1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311418AF532
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Human, Mouse, Bovine, Feline, Monkey samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
MUC5AC | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Bovine, Feline, Monkey | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
2-11M1 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient | |
0.1 mL | |
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
4586 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction