Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5AC Antibody (45M1), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP232732APCCY7
Description
MUC5AC Monoclonal specifically detects MUC5AC in Human, Mouse, Rat, Porcine, Chicken, Feline, Mammal, Monkey, Rabbit, Bovine (Negative) samples. It is validated for Flow Cytometry, Dot Blot.Specifications
MUC5AC | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MUC5AC | |
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient | |
0.1 mL | |
Primary | |
This monoclonal antibody recognizes the peptide core of gastric mucin M1 (recently identified as Mucin 5AC).Its epitope is located in the C-terminal cysteine rich part of the peptide core of MUC5AC. Its epitope is destroyed by beta-mercaptoethanol but not by periodate treatment. This mucin is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
45M1 | |
Flow Cytometry | |
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
Mouse | |
Protein A or G purified | |
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
4586.0 | |
Human, Mouse, Rat, Pig, Chicken, Feline, Primate, Rabbit, Bovine (Negative) | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction