Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC5AC Monoclonal Antibody (58M1)
GREENER_CHOICE

Catalog No. 01672013
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-013 Supplier Invitrogen™ Supplier No. MA135707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

58M1 recognizes the peptide core of gastric mucin M1 (now: MUC5AC), and more specifically with the e epitope amongst the a, b, c, d, e, f, g and h protein core epitopes defined by Bara for M1. MUC5AC is present in primary ovarian mucinous cancer and gastric cancer, but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification pancreatic carcinoma and pre-cancerous changes vs. normal pancreas.

The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC5AC
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 58M1
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MUC5AC
Gene Accession No. P98088
Gene Alias 2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Gene Symbols MUC5AC
Host Species Mouse
Immunogen Mucin isolated from an ovarian cyst fluid.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100847395, 105261014, 17833, 4586
Target Species Human, Mouse, Monkey, Bovine, Feline
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.