Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC5AC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579705
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579705 Supplier Invitrogen™ Supplier No. PA579705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: human gastric carcinoma tissue, rat stomach tissue.

The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC5AC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene MUC5AC
Gene Accession No. P98088
Gene Alias 2210005L13Rik; gastric mucin; LeB; lewis B blood group antigen; LOW QUALITY PROTEIN: mucin-5AC; major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Gene Symbols MUC5AC
Host Species Rabbit
Immunogen E.coli-derived human Mucin 5AC recombinant protein (Position: D4848-H5030).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4586, 682837
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.