Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC5AC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987760
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7760 Supplier Bioss Supplier No. BS7166R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.

Specifications

Antigen MUC5AC
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MUC5AC
Gene Accession No. P98088
Gene Alias 2210005L13Rik; Gastric mucin; LeB; Lewis B blood group antigen; Major airway glycoprotein; MGM; MUC5; MUC5AC; MUC-5AC; mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5AC, oligomeric mucus/gel-forming; mucin 5AC, oligomeric mucus/gel-forming pseudogene; Mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Gene Symbols MUC5AC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MUC5AC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4586
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.