Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5B Antibody (19.4E), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229706AF647
Description
MUC5B Monoclonal antibody specifically detects MUC5B in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
MUC5B | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Primate, Mouse (Negative) | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunohistochemistry | |
19.4E | |
Western Blot, ELISA, Immunohistochemistry | |
high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial | |
Un-glycosylated, serine- and threonine-free synthetic peptide from the N-terminus of human MUC5B. | |
0.1 mL | |
Cell Cycle and Replication, Cellular Signaling | |
727897 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction