Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5B Antibody (4A10-H2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250522F
Description
MUC5B Monoclonal specifically detects MUC5B in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Mucin 5B | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MUC5B | |
KLH conjugated peptide RNREQVGKF in 4 of the cysteine domains of the tandem repeat region was used to immunise Balb/c female mice. | |
0.1 mL | |
Cell Cycle and Replication, Cellular Signaling | |
727897 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
4A10-H2 | |
ELISA, Immunohistochemistry | |
high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction