Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC5B Monoclonal Antibody (C10)
GREENER_CHOICE

Catalog No. PIMA541641
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541641 200 μL
PIMA543650 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541641 Supplier Invitrogen™ Supplier No. MA541641
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MUC5B Monoclonal Antibody for Western Blot

Mucins (MUCs) are heavily glycosylated glycoproteins that are synthesized either as membrane-bound or secreted proteins. Mucins are noteworthy because they have an aberrant expression profile in various malignancies. Mucins normally play a protective role for epithelial tissues and are expressed in the gastrointestinal, respiratory, and genito-urinary tracts. Mucins may also be involved in the renewal and differentiation of the epithelium, and the modulation of cell adhesion and cell signaling. MUC5B is thought to play a vital role in maintaining oral health, and may also be involved in respiratory and oral disease.2 MUC5B expression has been studied in gastric carcinoma as well as chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC5B
Applications Western Blot
Classification Monoclonal
Clone C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MUC5B
Gene Accession No. Q9HC84
Gene Alias Cervical mucin; cervical mucin MUC5B; high molecular weight salivary mucin MG1; MG1; MUC5; MUC5B; MUC-5B; MUC9; mucin 5, subtype B, tracheobronchial; mucin 5B, oligomeric mucus/gel-forming; Mucin-5 subtype B, tracheobronchial; Mucin-5B; sublingual gland mucin
Gene Symbols MUC5B
Host Species Mouse
Immunogen Recombinant protein Mucin 5 Subtype B. The antigen corresponds to amino acid range 5653-5762 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.