Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC6 Antibody (CLH5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311450PE
Description
MUC6 Monoclonal antibody specifically detects MUC6 in Human samples. It is validated for Flow CytometrySpecifications
MUC6 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
4588 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
CLH5 | |
Flow Cytometry | |
gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6 | |
A synthetic peptide of the Gastric MUC6 tandem repeat sequence (Uniprot: Q6W4X9 ) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction