Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550557 Supplier Invitrogen™ Supplier No. PA550557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MUC6 protein.

Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MUC6
Gene Accession No. Q6W4X9
Gene Alias Gastric mucin-6; gastric mucin-like protein; MUC6; MUC-6; mucin 6; mucin 6, gastric; mucin 6, oligomeric mucus/gel-forming; mucin-6; secreted gel-forming mucin-6
Gene Symbols MUC6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human mucin 6, oligomeric mucus/gel-forming.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4588
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.