Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552292
Description
Immunogen sequence: RRHHHQSPKS HFELPHYPGL LAHQKPFIRK SYKCLHKRCR PKLPPSPNNP PKFPNPHQPP KHPDKNSSVV NPTLVATTQI PSVTFPSAST KITTLPNVTF LPQNATTISS RENVNTSSSV ATLAPVNSPA P Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.
This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. This antimicrobial protein has antibacterial and antifungal activity. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
Specifications
MUC7 | |
Polyclonal | |
Unconjugated | |
MUC7 | |
apo-MG2; MG2; MUC7; MUC-7; mucin 7, salivary; mucin 7, secreted; mucin-7; Salivary mucin-7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAX7 | |
MUC7 | |
Recombinant protein corresponding to Human MUC7. Recombinant protein control fragment (Product #RP-88569). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction