Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579707
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SW620 whole cell, human HepG2 whole cell.
This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. This antimicrobial protein has antibacterial and antifungal activity. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
Specifications
MUC7 | |
Polyclonal | |
Unconjugated | |
MUC7 | |
apo-MG2; MG2; MUC7; MUC-7; mucin 7, salivary; mucin 7, secreted; mucin-7; Salivary mucin-7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4589 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
Q8TAX7 | |
MUC7 | |
A synthetic peptide corresponding to a sequence of human MUC7 (QNKISRFLLYMKNLLNRIIDDMVEQ). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction