Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mucin 4X/Mucin 4Y Recombinant Rabbit Monoclonal Antibody (HL2026)
GREENER_CHOICE

Catalog No. FEMA550363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550363 Supplier Invitrogen™ Supplier No. MA550363
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Mucin 4X/Mucin 4Y Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Mucin 4X/Mucin 4Y (MUC4) is a gene target that codes for a transmembrane glycoprotein expressed in various epithelial tissues. MUC4 plays an important role in cellular adhesion, signaling, and protection of the epithelial surface. It is often overexpressed in various cancers, including pancreatic, ovarian, and breast cancers, and has been identified as a potential biomarker for these cancers. Targeting MUC4 has shown promise as a therapeutic strategy for cancer treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mucin 4X/Mucin 4Y
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2026
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MUC4
Gene Accession No. Q99102
Gene Alias Mucin-4X/Mucin-4Y
Gene Symbols MUC4
Host Species Rabbit
Immunogen Recombinant fragment of human Mucin 4 X+Mucin 4 Y.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4585
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.