Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mucin 6 (MUC-6) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1153R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1153R7 7 mL Prediluted
MS1153S0 100 μL Concentrated
MS1153S1 500 μL Concentrated
MS1153S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1153R7 Supplier Epredia™ Supplier No. MS1153R7

Deliver accurate, reproducible results with the Epredia™ Mucin 6 (MUC-6) Ab-1, Mouse Monoclonal Antibody.

Mucin 6 (Muc-6 Glycoprotein). Mucins are heavily glycosylated glycoproteins, which constitute the major components of mucus which cover the surface of epithelial tissues. Nine distinct epithelial mucin genes (Muc-1, 2, 3, 4, 5AC, 5B, 6, 7 and 8) have been identified. Various immunohistochemical and in situ hybridization studies have shown that these mucins are differentially expressed in epithelia with cell-type specificity. Muc-5AC and Muc-6 glycoproteins are the first two mucins found in superficial epithelium and Muc-6 glycoprotein is found in the deep glands.

Host Species: Mouse

Clone: CLH5

Isotype: IgG1

Species Reactivity: Human. Weakly react with Rat. Others not known.

Epitope: Not determined

Immunogen: Synthetic peptide of the tandem repeat sequence of human MUC-6 protein

Positive Control: Stomach

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Mucin 6 (MUC-6) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CLH5
Conjugate Unconjugated
Host Species Mouse
Immunogen Synthetic peptide of the tandem repeat sequence of human Muc-6 protein
Purification Method Stomach
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.