Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mucin, Bovine Submaxillary Gland, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
Description
General Description
- Mucin derived from the bovine submaxillary gland is a glycoprotein present in saliva, gastrointestinal juices, and other secretions that promote cilia lubrication
Application
- Mucin can be used as a source of O-linked oligosaccharides for the viral enzyme, neuraminidase
- Mucins are glycoproteins with roles in immunity and digestion
- In biological research, mucin is used to investigate a galactose-specific lectin from the red marine alga Ptilota filicina
- It is used to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line
- It is used as a substrate for recombinant sialidase Gardnerella vaginalis
- It is used in protocols to quantify mucin

Specifications
Specifications
For Use With (Application) | Neuraminidase Substrate |
Formulation | Lyophilized Powder |
Quantity | 500 mg |
Source | Bovine Submaxillary Gland |
Storage Requirements | 2° to 8°C |
Regulatory Status | RUO |
Product Type | Mucin |
Recombinant | Native |
Form | Lyophilized |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction