Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mucolipin 1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11082375AF405
Description
Mucolipin 1 Polyclonal specifically detects Mucolipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Mucolipin 1 | |
Polyclonal | |
Western Blot | |
MG-2, ML4TRP-ML1, MLIV, MST080, MSTP080, mucolipidin, mucolipidosis type IV protein, mucolipin 1, mucolipin-1, TRPML1, TRP-ML1, TRPM-L1 | |
Rabbit | |
66 kDa | |
0.1 mL | |
Cell Biology, Neuroscience, Sensory Systems, Signal Transduction, Vision | |
57192 | |
Store at 4C in the dark. |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MCOLN1 | |
Synthetic peptide made to a C-terminal portion of mouse Mucolipin 1 (within residues 500-580). [Swiss-Prot# Q99J21] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction