Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP5M1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548964
Description
Predicted to react with monkey based on sequence homology.
As part of AP-5, a probable fifth adaptor protein complex it may be involved in endosomal transport. According to PubMed:18395520, it may play a role in cell death.
Specifications
AP5M1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H0R1 | |
AP5M1 | |
KLH conjugated synthetic peptide between 356-384 amino acids from the C-terminal region of human MUDEN. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
AP5M1 | |
AP-5 complex subunit mu-1, Adaptor-related protein complex 5 subunit mu-1, Mu5, Mu-2-related death-inducing protein, MuD, Putative HIV-1 infection-related protein, AP5M1, C14orf108, MUDENG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55745 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction