Learn More
Thermo Scientific™ Mueller Hinton Agar
Description
The Thermo Scientific™ Remel Mueller Hinton Agar is used for antimicrobial disk diffusion susceptibility testing.
Specifications
Specifications
| Certifications/Compliance | Conforms to CLSI standards. Industrial Reference: BAM, COMPF. |
| Description | Mueller Hinton Agar |
| Form | Powder |
| Media Type | Mueller Hinton Agar |
| Product Type | Nutrient Agar |
| Quantity | 2.5 kg |