Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mueller Hinton Agar

Catalog No. r01624
Click to view available options
Quantity:
10/Pk.
100/Cs.
40/Pk.
Format:
15 x 100mm Plate
15 x 150mm Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.

The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF.
Description Mueller Hinton Agar
Format 15 x 100mm Plate
Manufacturing Origin United States
Product Type Agar
Quantity 100/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.