Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MuERVL-Gag Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP266963
Description
MuERVL-Gag Polyclonal specifically detects MuERVL-Gag in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MuERVL-Gag | |
Polyclonal | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
Rabbit | |
Immunogen affinity purified | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Immunocytochemistry/Immunofluorescence 1:200 | |
Gag polyprotein | |
Synthetic peptide within human MuERVL-Gag aa 541-581/581. (SwissProt: V9H130 Mouse) | |
100 μL | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction