Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MuERVL-Gag Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119739 Supplier Invitrogen™ Supplier No. PA5119739
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse embryo.

A large number of retrotransposons are expressed when the zygotic genome is first transcribed, including the endogenous retroviruses (ERVs), LINE-1 elements, and the non-autonomous SINE elements. At the 2C stage, MuERV-L/MERVL retrovirus-like elements are transiently de-repressed and produce 3% of the transcribed mRNAs. Following the 2C stage, MERVL-retroelement expression is silenced. The newly study discovered that this regulated pattern of MERVL expression overlapped with greater than one hundred 2C-specific genes that have co-opted regulatory elements from these foreign retroviruses to initiate their transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MuERVL-Gag
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias Murine Endogenous Retrovirus-Gag
Host Species Rabbit
Immunogen Synthetic peptide within mouse MuERVL-Gag aa 541-581.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.