Learn More
Invitrogen™ MUL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113767
Description
Immunogen sequence: GMQYYLSSQD FDSLLQRQES SVRLWKVLAL VFGFATCATL FFILRKQYLQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
E3 ubiquitin-protein ligase that plays a role in the control of mitochondrial morphology. Promotes mitochondrial fragmentation and influences mitochondrial localization. Inhibits cell growth. When overexpressed, activates JNK through MAP3K7/TAK1 and induces caspase-dependent apoptosis. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
Specifications
| MUL1 | |
| Polyclonal | |
| Unconjugated | |
| MUL1 | |
| 0610009K11Rik; AV000801; C1orf166; E3 SUMO-protein ligase MUL1; E3 ubiquitin ligase; E3 ubiquitin-protein ligase MUL1; Gide; growth inhibition and death E3 ligase; MAPL; mitochondria-anchored protein ligase; mitochondrial E3 ubiquitin ligase 1; mitochondrial E3 ubiquitin protein ligase 1; mitochondrial ubiquitin ligase activator of NF-kB; mitochondrial ubiquitin ligase activator of NFKB 1; Mitochondrial-anchored protein ligase; MUL1; MULAN; Putative NF-kappa-B-activating protein 266; RGD1309944; ring finger protein 218; RING-type E3 ubiquitin transferase NFKB 1; RNF218; RP11-401M16.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79594 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q969V5 | |
| MUL1 | |
| Synthetic peptide directed towards the middle region of human C1orf166. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.