Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988962
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8962 Supplier Bioss Supplier No. BS9291R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E3 ubiquitin-protein ligase that plays a role in the control of mitochondrial morphology. Promotes mitochondrial fragmentation and influences mitochondrial localization. Inhibits cell growth. When overexpressed, activates JNK through MAP3K7/TAK1 and induces caspase-dependent apoptosis. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.

Specifications

Antigen MUL1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MUL1
Gene Accession No. Q969V5
Gene Alias 0610009K11Rik; AV000801; BOS_2439; C1orf166; E3 SUMO-protein ligase MUL1; E3 ubiquitin ligase; E3 ubiquitin-protein ligase MUL1; GIDE; Growth inhibition and death E3 ligase; MAPL; mitochondria-anchored protein ligase; mitochondrial E3 ubiquitin ligase 1; mitochondrial E3 ubiquitin protein ligase 1; mitochondrial ubiquitin ligase activator of NF-kB; Mitochondrial ubiquitin ligase activator of NFKB 1; Mitochondrial-anchored protein ligase; MUL1; MULAN; Putative NF-kappa-B-activating protein 266; RGD1309944; RING finger protein 218; RING-type E3 ubiquitin transferase NFKB 1; RNF218; RP11-401M16.2
Gene Symbols MUL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MUL1/RNF218.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79594
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.