Learn More
Invitrogen™ MULT1 (NKG2D Ligand) Polyclonal Antibody

Description
Positive Samples: Raji; Cellular Location: Cell membrane, Endoplasmic reticulum, GPI-anchor, Lipid-anchor.
Specifications
Specifications
| Antigen | MULT1 (NKG2D Ligand) |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 7.06 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ULBP1 |
| Gene Accession No. | Q9BZM6 |
| Gene Alias | A430108B07Rik; ALCAN-beta; MULT1; N2DL1; N2DL-1; NKG2D ligand; NKG2D ligand 1; NKG2D ligand1; NKG2DL1; RAET1I; retinoic acid early transcript 1I; UL16 binding protein 1; UL16-binding protein 1; UL16-binding protein-like transcript 1; ULBP; ULBP 1; ULBP1 |
| Gene Symbols | ULBP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.