Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MULT1, Rat anti-Mouse, Clone: 1D6, Millipore Sigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABS1906100UG
Description
Anti-MULT1, clone 1D6, Cat. No. MABS1906, is a rat monoclonal antibody that detects UL16-binding protein-like transcript 1 splicing variant and has been tested for use in ELISA, Flow Cytometry, and Immunocytochemistry.
UL16-binding protein-like transcript 1 splicing variant (UniProt: Q330P3; also known as MULT1, MULT-1, ULBP1) is encoded by the Ulbp1 gene in murine species. MULT-1 is a MHC Class I-like molecule that serves as a high affinity ligand for natural Killer Group 2D (NKG2D) receptor. It is a type I transmembrane protein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form. Its transmembrane domain is localized to amino acids 124 to 146 and its extracellular domain contains an alpha 1 and alpha 2 like domain with two intrachain disulfide bonds. MULT-1 expression is reported to be low or absent in normal adult tissues, however, under conditions of stress and in tumor cells its levels are elevated. Many tumor cells are shown to release soluble NKG2D ligands through proteolytic shedding, alternative splicing, or exosome secretion. In mice MULT-1 is shown to cause NK cell activation and tumor rejection. Mice subjected to cytomegalovirus (MCMV) infection display strong expression of Ulbp1 (Mult1) gene, but surface expression of MULT-1 is nevertheless completely abolished by the virus. Fc receptor fcr-1 is reported to down-regulate MULT-1 levels by interfering with surface MULT-1 and causing its subsequent degradation in lysosomes. This effect of Fcr-1 can be completely abolished by imipramine (Cat. No. I0899), which blocks AP2-mediated clathrin pathway. (Ref.: Lenac, T., et al. (2006). J. Exp. Med. 203(8); 1843 1850; Krmpotic, A., et al. (2005). J. Exp. Med. 201(2): 211-220; Deng, W., et al. (2015). Science. 348(6230); 136-139).
Specifications
MULT1 | |
Monoclonal | |
Unconjugated | |
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Ulbp1 | |
Recombinant fragment corresponding to 186 amino acids from the extracellular domain of murine UL16-binding protein-like transcript 1 splicing variant (MULT-1). | |
100 μg | |
Signaling | |
NA | |
Mouse | |
Purified |
ELISA, Flow Cytometry, Immunocytochemistry | |
1D6 | |
Immunocytochemistry Analysis: A representative lot detected MULT1 in Immunocytochemistry applications (Lenac, T., et. al. (2006). J Exp Med. 203(8):1843-50; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20).Flow Cytometry Analysis: A representative lot detected MULT1 in Flow Cytometry applications (Lenac, T., et. al. (2006). J Exp Med. 203(8):1843-50; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20).ELISA Analysis: A representative lot detected MULT1 in ELISA applications (Sheppard, S., et. al. (2017). Nat Commun. 8:13930; Krmpotic, A., et. al. (2005). J Exp Med. 201(2):211-20). | |
MULT-1;ULBP1;mouse UL16-binding protein-like Transcript 1;NKG2DL;NKG2D ligand | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Clone 1D6 is a rat monoclonal antibody that detects UL16-binding protein-like transcript 1 splicing variant in mouse. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction