Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Element Organometallic Oil Standards 23 Element, 100μg/g; SPEX CertiPrep™
SDP

Catalog No. s23100y
Click to view available options
Quantity:
50 g
100 g

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/g
Al100μg/g
B100μg/g
Ba100μg/g
Ca100μg/g
Cd100μg/g
Cr100μg/g
Cu100μg/g
Fe100μg/g
K100μg/g
Mg100μg/g
Mn100μg/g
Mo100μg/g
Na100μg/g
Ni100μg/g
P100μg/g
Pb100μg/g
Sb100μg/g
Si100μg/g
Sn100μg/g
Ti100μg/g
V100μg/g
Zn100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 23 Element Oil Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al,Ba,B
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, Ba, B
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.