Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Power Adapters for Digital Vortex Mixers

Power adapters for Digital Vortex Mixers

$35.35 - $53.90


Products 2
Catalog Number Mfr. No. Description Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Description Plug Type Price Quantity & Availability  
11-676-451
View Documents
Thermo Scientific™
88870124
Multi-power adapter 220V EU, UK, CN Plug
Each for $53.90
Add to cart
 
11-676-452
View Documents
Thermo Scientific™
88870125
Power adapter 120V US Plug
Each for $35.35
Add to cart
 
Description

Description

Power adapters for Digital Vortex Mixers, which offer accurate and efficient sample mixing

Service Plan

Standard Thermo warranty guideline, provide repair service and free spare part within warranty

Warranty and Services

2 years

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit