Learn More
Specifications
Specifications
| Assay Range | 1 - 300 ng/ml (0.1 - 30 ng/well) |
| Assay Sensitivity | 3 ng/ml |
| Product Type | ELISA Kit (Colorimetric) |
| Specificity | The dsRNA Detection Kit allows sensitive and selective detection of dsRNA molecules (larger than 30-40 bp), independent of their nucleotide composition and sequence. |
| For Use With (Application) | ELISA |
| Quantity | 200 Tests |
| Storage Requirements | Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Synonym | Double-stranded RNA |
| Target | Multi-Species |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
