Learn More
IKA™ Multi-use Milling Tube MMT 40.1
SDP
Catalog No. 01183085 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
40 mL
Catalog No. Capacity (Metric)
01-183-085 40 mL
1 options
This item is not returnable. View return policy
Catalog No. 01-183-085 Supplier IKA™ Supplier No. 20003165
Get a Magnetic Stirrer with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Durable, dishwasher-safe grinding chamber/milling tube.
  • Can be used and cleaned in a dishwasher multiple times
  • Package includes a rich set of spare parts so that wearing parts can be replaced if necessary
  • Delivery: 1 x MMT 40, 25 x sealing, 5 x beater, 5 x coupling
  • Additional abrasion set A-MMT 40.100 available
Specifications
Product Type Milling Tube, Grinding Chamber
Capacity (Metric) 40 mL
Includes Rich set of spare parts, 25 x sealings, 5 x beaters, 5 x couplings