Learn More
Dentec Safety™ Multi-Vapour Gas Cartridge
Dentec Safety™ Multi-Vapour Gas Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity.
$71.10 - $1047.00
Specifications
Material | Plastic |
---|---|
Product Type | Respirator Accessories |
For Use With (Equipment) | Masks |
Includes | Cartridges and filters |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-222-515
![]() |
Dentec Safety™
158T27 |
6/Box |
Pack of 6 for $71.10
|
|
|||||
19-222-516
![]() |
Dentec Safety™
158T27B100 |
100/Box |
Case of 100 for $1,047.00
|
|
|||||
Description
- Offset connector allows the user to rotate the cartridge into 3 different positions to improves visibility and allow face shields or welding helmets to be worn without interference
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Plastic | |
Respirator Accessories | |
Masks | |
Cartridges and filters |