Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKA™ MultiDrive TC1 Protective Housing
SDP

Catalog No. 01183110 Shop All IKA Works Products
Change view
Click to view available options
For Use With (Equipment):
IKA MultiDrive Control in combination with MT150 Milling Tube or BT250 Blending Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-183-110 IKA MultiDrive Control in combination with MT150 Milling Tube or BT250 Blending Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-183-110 Supplier IKA™ Supplier No. 20023489
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MultiDrive TC1 Protective Housing is a accessory for MultiDrive MT150 and BT250 tubes and is mandatory for operation.

  • For safety reasons, MultiDrive TC1 Protective Housing is mandatory to operate MT150 and BT250 tubes (MultiDrive will not start if TC1 isn't attached)
  • Solvent resistant
  • Dishwasher safe

Specifications

Product Type Protective Housing for MultiDrive MT150 and BT250
For Use With (Equipment) IKA MultiDrive Control in combination with MT150 Milling Tube or BT250 Blending Tube
Weight (Metric) 0.365 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.