Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MultiKnit™ 76-400 Heavyweight Polycotton Gloves

Catalog No. 19501043 Shop All Ansell Products
Change view
Click to view available options
Size:
7
9
10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-043 9
19-501-042 7
19-501-044 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-501-043 Supplier Ansell Supplier No. 222179
This item is not returnable. View return policy

Durable gloves are constructed from a heavyweight, polycotton blend to provide general protection for a wide variety of applications.

  • String-knit contstruction from heavyweight polycotton material
  • Provides moderate heat protection from temperatures up to 200°F
  • Continous knit wrist cuff style
  • Launderable for extended useful life
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Construction String Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Material Polycotton
Size 9
Temperature (Metric) Max 200°F
For Use With (Application) Carpentry, Food Processing, Janitorial Services, Metal Fabrication, and Sanitation
Glove Cuff Style Knit
Liner Material Unlined
Series MultiKnit™ 76-400
Yarn Gauge 7
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.