Learn More
Cytiva Multiphor II Buffer Strip Positioner, complete
Catalog No. 45001721 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Multiphor II Cooling Plate
Catalog No. For Use With (Equipment)
45-001-721 Multiphor II Cooling Plate
1 options
Catalog No. 45-001-721 Supplier Cytiva Supplier No. 80644290
Multiphor II Buffer Strip Positioner is a frame with slots that sits on top of ExcelGel precast gel after the gel is placed on Multiphor II cooling plate.
The slots in the positioner facilitate placement of the buffer strips for electrophoresis and hold the strips securely in place during the run.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Cooling Plate
Type Multiphor II Buffer Strip Positioner