Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS Durapore™ 96-Well Filter Plates

Catalog No. MSHVN4B50 Shop All MilliporeSigma Products
Click to view available options
Pore Size:
0.22 μm
0.45 μm
0.65 μm
1.2 μm
Color:
Clear
Opaque
Material:
Barex
Styrene
Quantity:
10/Pk.
50/Pk.

Designed for full automation compatibility. MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Material Barex
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28 cm2
For Use With (Application) Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
Quantity 50/Pk.
Dimensions (L x W x H) 0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44 cm)
Length (Metric) 1.28 cm
Width (Metric) 8.55 cm
Height (Metric) 1.44 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.