Learn More
Siemens Healthineers Multistix™ and Uristix™ Reagent Strips
Catalog No. 23042305 Shop All Siemens Healthineers Products
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity
Glucose, Leukocyte, Nitrite, Protein
Description:
Multistix 10SG
Multistix 5
Multistix 7
Multistix 8 SG
Uristix 4
This item is not returnable. View return policy
This item is not returnable. View return policy
Visually and instrumentally readable urinalysis strips
  • Includes testing for a variety of parameters
  • Supplied in lightweight, unbreakable, plastic vials
  • Provide diagnostically useful information about the status of carbohydrate metabolism, kidney and liver function, acid-base balance and bacteriuria-pyuria
TRUSTED_SUSTAINABILITY
Specifications
Description Multistix 10SG
Detectable Analytes Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
For Use With (Application) Urinalysis
Quantity 100 Pack
Test Time 1 to 2 min.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.