Learn More
Siemens Healthineers Multistix™ and Uristix™ Reagent Strips
Shop All Siemens Healthineers Products
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity
Glucose, Leukocyte, Nitrite, Protein
Description:
Multistix 10SG
Multistix 5
Multistix 7
Multistix 8 SG
Uristix 4
Description
- Includes testing for a variety of parameters
- Supplied in lightweight, unbreakable, plastic vials
- Provide diagnostically useful information about the status of carbohydrate metabolism, kidney and liver function, acid-base balance and bacteriuria-pyuria
Specifications
Specifications
| Description | Multistix 10SG |
| Detectable Analytes | Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen |
| For Use With (Application) | Urinalysis |
| Quantity | 100 Pack |
| Test Time | 1 to 2 min. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov