Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MuLV TM Mouse anti-Virus, Clone: 42-114, MilliporeSigma™

Catalog No. MABF207425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2074-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2074-25 Supplier MilliporeSigma Supplier No. MABF207425UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MuLV TM, clone 42-114 is a mouse monoclonal antibody that detects Murine leukemia virus (MuLV) transmembrane protein and has been tested for use in Immunoprecipitation and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MuLV TM
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 42-114
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen AKR K36 and AKR SL3 leukemia virus.
Purification Method Purified by ion-exchange chromatography
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 42-114 detects murine leukemia virus in infected cells.
Target Species Virus
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.