Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUM1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178760AF647
Description
MUM1 Polyclonal antibody specifically detects MUM1 in Human samples. It is validated for Western BlotSpecifications
MUM1 | |
Polyclonal | |
Western Blot | |
EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
The immunogen for this product maps to a region between residue 125 and 175 of human Melanoma Associated Antigen (Mutated) 1 using the numbering given in entry NP_116242.2 (GeneID 84939). | |
0.1 mL | |
Melanoma Cell Markers | |
84939 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction