Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUM1 Mouse anti-Human, Clone: 3B1D2, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 603531IG150UL

Description
Mutated melanoma-associated antigen 1 (MUM1) is a 78 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. Antibodies to MUM1 are useful in the sub classification of lymphoid maligcies.Specifications
MUM1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2TAK8 | |
MUM1 | |
MUM1 Fusion Protein Ag3371 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
3B1D2 | |
Unconjugated | |
MUM1 | |
EXPAND1, HSPC211, MUM 1, MUM1, Protein expandere | |
Mouse | |
Protein A | |
RUO | |
84939 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction