Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUM1 Mouse anti-Human, Clone: MUM1p, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB2003560.125ML
This item is not returnable.
View return policy
Description
MUM1 Monoclonal specifically detects MUM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| MUM1 | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with sodium azide | |
| MUM1 | |
| Recombinant GST-MUM1 protein. | |
| 0.125 ml | |
| Melanoma Cell Markers | |
| 84939 | |
| Human | |
| Supernatant |
| Immunohistochemistry (Paraffin) | |
| MUM1p | |
| Immunohistochemistry 1:10-1:25, Immunohistochemistry-Paraffin 1:10-1:25, Immunohistochemistry-Frozen 1:10-1:500 | |
| EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Multiple myeloma oncogene-1 (MUM1) is a 50kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction