Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115603 Supplier Invitrogen™ Supplier No. PA5115603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MUM1.

Mutated melanoma-associated antigen 1 (MUM1) is a 78 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. Antibodies to MUM1 are useful in the sub classification of lymphoid malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MUM1
Gene Accession No. B1H224, Q2TAK8, Q6DID5
Gene Alias 2610019J09; 9430059D04Rik; EXPAND1; FLJ14868; FLJ22283; HSPC211; melanoma associated antigen (mutated) 1; melanoma ubiquitous mutated protein; MGC131891; MGC163315; Mum1; MUM-1; Mutated melanoma-associated antigen 1; protein expandere; PWWP domain-containing DNA repair factor 3A; PWWP domain-containing protein MUM1; Pwwp3a; UBE-1C2
Gene Symbols MUM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MUM1(Accession Q2TAK8), corresponding to amino acid residues I665-R710.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362838, 68114, 84939
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.