Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mumps Virus NP Monoclonal Antibody (6007)
GREENER_CHOICE

Catalog No. PIMA533281
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533281 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533281 Supplier Invitrogen™ Supplier No. MA533281
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Does not cross react with: Measles virus, Respiratory Syncytial virus, Parainfluenza 1, 2 or 3 viruses.

Mumps rubulavirus is the causative agent of mumps. The signs of mumps include swelling of the parotid glands, salivary glands and other epithelial tissues. Symptoms of mumps are fatigue, body aches, headache, loss of appetite, low grade fever, swelling of the salivary glands. Mumps can also result in muscle pain, deafness, meningitis, pancreatitis, swelling of testicles or ovaries, and death. Most people who contract mumps show symptoms of the virus, however there are few who show no or very few symptoms. Natural infection is currently restricted to humans and the virus is transmitted by direct contact, droplet spread, or contaminated objects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mumps Virus NP
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6007
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias mumps virus nuclear
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.