Learn More
Invitrogen™ Mumps Virus NP Monoclonal Antibody (6008)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547563
Description
Specificity: Specific for the nucleoprotein of Mumps virus (see below). Does not cross react with: Measles virus, Respiratory Syncytial virus, Parainfluenza 1, 2 or 3 viruses. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Mumps rubulavirus is the causative agent of mumps. The signs of mumps include swelling of the parotid glands, salivary glands and other epithelial tissues. Symptoms of mumps are fatigue, body aches, headache, loss of appetite, low grade fever, swelling of the salivary glands. Mumps can also result in muscle pain, deafness, meningitis, pancreatitis, swelling of testicles or ovaries, and death. Most people who contract mumps show symptoms of the virus, however there are few who show no or very few symptoms. Natural infection is currently restricted to humans and the virus is transmitted by direct contact, droplet spread, or contaminated objects.
Specifications
Mumps Virus NP | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry | |
6008 | |
Unconjugated | |
mumps virus nuclear | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.