Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Munc13-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589018 Supplier Invitrogen™ Supplier No. PA589018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIRKYFCSRI QQQAETTSEE LGAVTVKASY RASEQKLRVE LLSASSLLPL DSNGSSDPFV QLTLEPRHEF PELAARETQK HKKDLHPLFD ETFEFLVPAE PCRKAGACLL LTVLDYDTLG ADDLEGEAFL PLREVPGLSG SEEPGEVPQT RLPLTYPAPN GDPILQLLEG RKGDREAQVF VRLRRHRAKQ ASQHALRPAP; Positive Samples: A549, HeLa, HepG2, Mouse lung; Cellular Location: Cytoplasm, Late endosome, Lysosome, Membrane, Peripheral membrane protein, Recycling endosome.

This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Munc13-4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Unc13d
Gene Accession No. B2RUP2, Q70J99
Gene Alias 2610108D09Rik; FHL3; HLH3; HPLH3; Jinx; likely ortholog of H. sapiens unc-13 homolog D (UNC13D); mFLJ00067; munc 13-4; Munc13-4; Protein unc-13 homolog D; unc-13 homolog D; unc-13 homolog D (C. elegans); Unc13d
Gene Symbols Unc13d
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201294, 70450
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.