Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Munc13-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119767 Supplier Invitrogen™ Supplier No. PA5119767
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Jurkat cell lysate, K562 cell lysate, rat lung tissue lysate, Hela, SHSY5Y, rat spleen tissue, human placenta tissue. Subcellular Location: Cytoplasm, Endosome, Lysosome, Membrane.

This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Munc13-4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Unc13d
Gene Accession No. Q70J99, Q9R189
Gene Alias 2610108D09Rik; FHL3; HLH3; HPLH3; Jinx; likely ortholog of H. sapiens unc-13 homolog D (UNC13D); mFLJ00067; munc 13-4; Munc13-4; Protein unc-13 homolog D; unc-13 homolog D; unc-13 homolog D (C. elegans); Unc13d
Gene Symbols Unc13d
Host Species Rabbit
Immunogen Recombinant protein within human Munc13-4 aa 900-1090.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192177, 201294
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.