Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Munc13-4 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518550
Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
Specifications
Munc13-4 | |
Polyclonal | |
Unconjugated | |
Unc13d | |
2610108D09Rik; FHL3; HLH3; HPLH3; Jinx; likely ortholog of H. sapiens unc-13 homolog D (UNC13D); mFLJ00067; munc 13-4; Munc13-4; Protein unc-13 homolog D; unc-13 homolog D; unc-13 homolog D (C. elegans); Unc13d | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
201294, 70450 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
B2RUP2, Q70J99 | |
Unc13d | |
Synthetic peptide sequence (ETQKHKKDLHPLFD) corresponding to the internal amino acids of UNC13D (aa 967-980). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction