Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUNC18 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518212
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Specifications
MUNC18 | |
Polyclonal | |
Unconjugated | |
Stxbp1 | |
AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
6812 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P61764 | |
Stxbp1 | |
Synthetic peptide sequence (SRVSFEDQAPTME) corresponding to the C-terminus amino acids of STXBP1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction