Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUNC18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530184
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530184 Supplier Invitrogen™ Supplier No. PA530184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (93%), Chicken (98%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUNC18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Stxbp1
Gene Accession No. O08599, P61764, P61765
Gene Alias AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h
Gene Symbols Stxbp1
Host Species Rabbit
Immunogen Recombinant protein fragment corresponding to a region within amino acids 354 and 574 of Human STXBP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20910, 25558, 6812
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.