Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mup6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144230
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144230 Supplier Invitrogen™ Supplier No. PA5144230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mup6 Polyclonal Antibody for ICC/IF, ELISA

Binds pheromones that are released from drying urine of males. These pheromones affect the sexual behavior of females.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mup6
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Mup6
Gene Accession No. P02762
Gene Alias Alpha-2U-globulin; BS6; Group 1; Major urinary protein 6; MUP 6; Mup6; MUP6_MOUSE
Gene Symbols Mup6
Host Species Rabbit
Immunogen Recombinant Mouse Major urinary protein 6 protein (19-180AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100038948, 620807
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.